Learn More
Biotium Catenin, delta-1 (CTNND1) (pTyr96) (25a), CF647 conjugate, 0.1mg/mL
Supplier:  Biotium BNC470339100
Encompass_Preferred
Delta 1 catenin, also known as p120 catenin, is a tyrosine kinase substrate that is phosphorylated in Src transformed cells or in response to growth factor stimulation. Like beta- and gamma-catenin, p120 Catenin directly associates with the cytoplasmic C-terminus of E-Cadherin via its Arm domain and may similarly interact with other cadherins. The catenin/cadherin complexes play an important role mediating cellular adhesion, including adherens junctions.Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-252-4973