Learn More
Cell Signaling Technology Catenin Delta-1 Monoclonal Antibody, Rabbit IgG, 20 μL
SDP
Catalog No. 502732689
Encompass
Catenin delta-1 (D7S2M) Rabbit Monoclonal Antibody is a recombinant antibody that recognizes endogenous levels of total catenin δ-1 protein. This monoclonal antibody is produced by immunizing animals with a recombinant protein specific to the amino terminus of human catenin δ-1.
  • Recognizes endogenous levels of total catenin δ-1 protein
  • Tested in human, mouse and rat species
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and less than 0.02% sodium azide
  • Validated for Western blotting, immunoprecipitation, immunohistochemistry (paraffin) and immunofluorescence (immunocytochemistry)
  • Requires storage at −20°C; do not aliquot the antibody
Catalog No. 50-273-2689 Supplier Cell Signaling Technology Supplier No. 59854T
Catenin delta-1 (D7S2M) Rabbit Monoclonal Antibody is a recombinant antibody that recognizes endogenous levels of total catenin δ-1 protein. This monoclonal antibody is produced by immunizing animals with a recombinant protein specific to the amino terminus of human catenin δ-1.
  • Recognizes endogenous levels of total catenin δ-1 protein
  • Tested in human, mouse and rat species
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and less than 0.02% sodium azide
  • Validated for Western blotting, immunoprecipitation, immunohistochemistry (paraffin) and immunofluorescence (immunocytochemistry)
  • Requires storage at −20°C; do not aliquot the antibody