Learn More
Biotium Catenin, gamma (Cardiomyocyte Marker) (15F11), CF488A conjugate, 0.1mg/mL
Supplier: Biotium BNC881420100
Gamma catenin, also known as JUP or junction plakoglobin, is a member of the catenin family. The catenins (alpha, beta, gamma, and delta) are ubiquitously expressed, cytoplasmic proteins that associate with E-cadherin at cellular junctions. Catenin/cadherin complexes play an important role in mediating cellular adhesion, including adherens junctions.Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.