Learn More
Medchemexpress LLC β-catenin-IN-2 | 1458664-10-2 | 99.6% | C15H14FN3 | 10MG
SDP
Catalog No. 5000259759
Encompass_Preferred
β-catenin-IN-2 (Compound H1B1) is a small-molecule inhibitor of β-catenin intended for preclinical research, particularly studies of colorectal cancer and Wnt/β-catenin signaling. The compound is characterized by CAS 1458664-10-2, molecular formula C15H14FN3, molecular weight 255.29, and is supplied with analytical documentation for research use.
  • Potent β-catenin inhibitor suitable for cellular and biochemical assays.
  • Reported applicability in colorectal cancer research.
  • High purity (99.6%) for reliable experimental results.
  • Characterized with COA, HNMR, and LCMS data available.
  • Small-molecule properties: molecular weight 255.29, formula C15H14FN3.
Catalog No. 50-002-59759 Supplier Medchemexpress LLC Supplier No. HY13646410MG
β-catenin-IN-2 (Compound H1B1) is a small-molecule inhibitor of β-catenin intended for preclinical research, particularly studies of colorectal cancer and Wnt/β-catenin signaling. The compound is characterized by CAS 1458664-10-2, molecular formula C15H14FN3, molecular weight 255.29, and is supplied with analytical documentation for research use.
  • Potent β-catenin inhibitor suitable for cellular and biochemical assays.
  • Reported applicability in colorectal cancer research.
  • High purity (99.6%) for reliable experimental results.
  • Characterized with COA, HNMR, and LCMS data available.
  • Small-molecule properties: molecular weight 255.29, formula C15H14FN3.