Learn More
Med Vet International Catego for Cats Over 1.5 Pounds and 8 Weeks or Older (3 Dose x 12)
Catalog No. 502188605
Encompass_Preferred
DescriptionCatego for Cats treats and prevents flea and tick infestation on cats and kittens weighing over 1.5 lbs and over 8 weeks of age. Active ingredients include: Dinotefuran 22.00%, Fipronil 8.92% and Pyriproxyfen 3.00%.*Kills adult fleas, flea eggs, flea larvae, ticks and chewing lice*Kills fleas within 6 hours*Kills ticks through contact so they do not have to bite to die*For both indoor and outdoor cats and kittensProducts are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-218-8605 Supplier Med Vet International Supplier No. PHCATEGO
DescriptionCatego for Cats treats and prevents flea and tick infestation on cats and kittens weighing over 1.5 lbs and over 8 weeks of age. Active ingredients include: Dinotefuran 22.00%, Fipronil 8.92% and Pyriproxyfen 3.00%.*Kills adult fleas, flea eggs, flea larvae, ticks and chewing lice*Kills fleas within 6 hours*Kills ticks through contact so they do not have to bite to die*For both indoor and outdoor cats and kittensProducts are not always manufacturer specific. Actual product appearance may differ from website image.