Learn More
Med Vet International Catgut Cassette #4/0 4/0 USP 50m(164')L, J0091C40
Catalog No. 5000582429
Encompass_Preferred
Catgut Medium Chromic. Manufactured under continuous supervision from selected materials processed to high quality standards. It is accurately gauged and evenly chromed. High knot-pull tensile strength. Good knot-holding ability and exceptionally good han
Catalog No. 50-005-82429 Supplier Med Vet International Supplier No. JOR91C40
Catgut Medium Chromic. Manufactured under continuous supervision from selected materials processed to high quality standards. It is accurately gauged and evenly chromed. High knot-pull tensile strength. Good knot-holding ability and exceptionally good han