Learn More
Carbosynth LLC. CATH-2 (Chicken), 5mg, MF= C147H245N51O30
SDP
Catalog No. 501952399
Encompass_Preferred
Chicken Cathelicidin-2; H-Arg-Phe-Gly-Arg-Phe-Leu-Arg-Lys-Ile-Arg-Arg-Phe-Arg-Pro-Lys-Val-Thr-Ile-Thr-Ile-Gln-Gly-Ser-Ala-Arg-Phe-NH2, Application: Antimicrobial Effects, Notes: Chicken cathelicidin-2 (CATH-2) belongs to a host defense peptide (HDP) family that has broad antimicrobial effects against gram-negative bacteria, gram-positive bacteria, and fungi, making it a potential antibiotic., salt: Trifluoroacetate Salt, one letter code: H-RFGRFLRKIRRFRPKVTITIQGSARF-NH2, MW: 3206.92
Catalog No. 50-195-2399 Supplier Carbosynth LLC. Supplier No. CAT3045PI5MG
Chicken Cathelicidin-2; H-Arg-Phe-Gly-Arg-Phe-Leu-Arg-Lys-Ile-Arg-Arg-Phe-Arg-Pro-Lys-Val-Thr-Ile-Thr-Ile-Gln-Gly-Ser-Ala-Arg-Phe-NH2, Application: Antimicrobial Effects, Notes: Chicken cathelicidin-2 (CATH-2) belongs to a host defense peptide (HDP) family that has broad antimicrobial effects against gram-negative bacteria, gram-positive bacteria, and fungi, making it a potential antibiotic., salt: Trifluoroacetate Salt, one letter code: H-RFGRFLRKIRRFRPKVTITIQGSARF-NH2, MW: 3206.92