Learn More
Med Vet International Stayfix Catheter Fixation Device, Large, 12 to 22 Fr., 685ME, 20/Box
Catalog No. 503045009
Encompass_Preferred
Special StayFix design secures catheters in place without the use of sutures, reducing the exposure to sharps and sharps-related accidents Provides safe and secure fixation for up to 7 days StayFix can be applied and removed without assitance Water resistant; can be worn in the shower Soft material conforms to the body and reduces catheter movement Hydrocolloid material reduces skin iritation by absorbing fluid leaking from the insertion site.
Catalog No. 50-304-5009 Supplier Med Vet International Supplier No. 721441BX
Special StayFix design secures catheters in place without the use of sutures, reducing the exposure to sharps and sharps-related accidents Provides safe and secure fixation for up to 7 days StayFix can be applied and removed without assitance Water resistant; can be worn in the shower Soft material conforms to the body and reduces catheter movement Hydrocolloid material reduces skin iritation by absorbing fluid leaking from the insertion site.