Learn More
Med Vet International Foley Catheter 30Cc Xx 18Fr, Each
Catalog No. 502830719
Encompass_Preferred
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. 2-way Foley. Sterile.
Catalog No. 50-283-0719 Supplier Med Vet International Supplier No. FC30X18
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. 2-way Foley. Sterile.