Learn More
Med Vet International Foley Catheter 30Cc X 30Fr, Each
Catalog No. 502830720
Encompass_Preferred
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. Two-way Foley. Sterile.
Catalog No. 50-283-0720 Supplier Med Vet International Supplier No. FC30X30
Add to Cart
Add to Cart
An innovative manufacturing process provides the maximum amount of silicone coating available. Latex catheter coated internally and externally in 100% silicone to resist encrustation and bacterial colonization. Two-way Foley. Sterile.