Learn More
Med Vet International Canine Urinary Catheter, 8Fr X 20", Sterile, J0154M, Each
Catalog No. 502832064
Encompass_Preferred
Sterile canine urinary catheter size 8fr X 50cm (20"). Constructed of a flexible transparent polypropylene, producing firmness without rigidity. Female luer fitting. Closed end with two lateral eyes.
Catalog No. 50-283-2064 Supplier Med Vet International Supplier No. JOR154M
Sterile canine urinary catheter size 8fr X 50cm (20"). Constructed of a flexible transparent polypropylene, producing firmness without rigidity. Female luer fitting. Closed end with two lateral eyes.