Learn More
Med Vet International Cath-Secure™ Holder, Tube, 5445-2, 50/Box
Catalog No. 503043038
Encompass_Preferred
Cath-Secure is the original multi-purpose tube holder. It provides an efficient, standardized system for securing catheters and medical tubing. The device is easy to adjust and may remain in place for approximately one week without altering skin integrity. Cath-Secure is made with alcohol-soluble adhesive
Catalog No. 50-304-3038 Supplier Med Vet International Supplier No. 192067BX
Cath-Secure is the original multi-purpose tube holder. It provides an efficient, standardized system for securing catheters and medical tubing. The device is easy to adjust and may remain in place for approximately one week without altering skin integrity. Cath-Secure is made with alcohol-soluble adhesive