Learn More
Med Vet International Cath-Secure™ Holder, Tube, 5445-2, Each
Catalog No. 503043039
Encompass_Preferred
Cath-Secure is the original multi-purpose tube holder. It provides an efficient, standardized system for securing catheters and medical tubing. The device is easy to adjust and may remain in place for approximately one week without altering skin integrity. Cath-Secure is made with alcohol-soluble adhesive
Catalog No. 50-304-3039 Supplier Med Vet International Supplier No. 192067EA
Cath-Secure is the original multi-purpose tube holder. It provides an efficient, standardized system for securing catheters and medical tubing. The device is easy to adjust and may remain in place for approximately one week without altering skin integrity. Cath-Secure is made with alcohol-soluble adhesive