Learn More
Med Vet International Dynarex Single Suction Catheter, Graduated w/Control Valve, 8 Fr, Pediatric, 50/Case
Catalog No. 502831939
Encompass_Preferred
Dynarex Resp-02™ Single Suction Catheters are designed to safely and effectively remove secretions from the respiratory tract of tracheostomy patients. The soft, flexible catheter with beveled tip is gentle on delicate mucosal tissue yet firm enough to allow for catheter rotation. The Catheter includes a vented cap port for enhanced suction control and two staggered eyelet openings to increase suctioning effectiveness and reduce patient trauma. Soft, pliable catheter with beveled tip provides optimal patient comfort Vented cap with control valve allows for greater procedural control Catheter tip is designed with 2 staggered eyelet openings for maximum suctioning efficiency Catheter features easy-to-read depth-number markings Coil packed for compact storage Sterile, single-patient use Not made with natural rubber latex or DEHP
Catalog No. 50-283-1939 Supplier Med Vet International Supplier No. 32001
Dynarex Resp-02™ Single Suction Catheters are designed to safely and effectively remove secretions from the respiratory tract of tracheostomy patients. The soft, flexible catheter with beveled tip is gentle on delicate mucosal tissue yet firm enough to allow for catheter rotation. The Catheter includes a vented cap port for enhanced suction control and two staggered eyelet openings to increase suctioning effectiveness and reduce patient trauma. Soft, pliable catheter with beveled tip provides optimal patient comfort Vented cap with control valve allows for greater procedural control Catheter tip is designed with 2 staggered eyelet openings for maximum suctioning efficiency Catheter features easy-to-read depth-number markings Coil packed for compact storage Sterile, single-patient use Not made with natural rubber latex or DEHP