Learn More
Med Vet International Slippery Sam, Tomcat Urethral Cather, No Side Holes, Open End, 3.5Fr X 4.0", Each
Catalog No. 502829902
Encompass_Preferred
A PTFE catheter with endhole for the drainage or administration of fluids Highly lubricious catheter shaft material ensures ease of catheter placement Low-profile silicone hub enables syringe attachment for flushing or connection to a closed collection system Suture holes in the silicone hub allow securing of the catheter to the prepuce Intended for one-time use Size: 3.5 French x 4"
Catalog No. 50-282-9902 Supplier Med Vet International Supplier No. SSTCATH3.5
 Restricted Item
Add to Cart
Add to Cart
A PTFE catheter with endhole for the drainage or administration of fluids Highly lubricious catheter shaft material ensures ease of catheter placement Low-profile silicone hub enables syringe attachment for flushing or connection to a closed collection system Suture holes in the silicone hub allow securing of the catheter to the prepuce Intended for one-time use Size: 3.5 French x 4"