Learn More
Med Vet International Catheter Insertion Tray Bardia Foley Without Catheter Without Balloon Without Catheter, 802010, Each
Catalog No. 503044788
Encompass_Preferred
(1) Inflation Syringe, 10 cc / Fenestrated Drape / Waterproof Underpad (3) Povidone-Iodine Swabsticks (1) Lubricating Jelly Packet, 5 grams (2) Gloves (1) Specimen Container with Label / Peel Top Package
Catalog No. 50-304-4788 Supplier Med Vet International Supplier No. 147913EA
 Restricted Item
Add to Cart
Add to Cart
(1) Inflation Syringe, 10 cc / Fenestrated Drape / Waterproof Underpad (3) Povidone-Iodine Swabsticks (1) Lubricating Jelly Packet, 5 grams (2) Gloves (1) Specimen Container with Label / Peel Top Package