Learn More
Med Vet International 12Fr Urethral Catheter, Plastic, Sterile
Catalog No. 502830799
Encompass_Preferred
Clear Vinyl Intermittent Self Catheters Suitable for male or female catheterization. Smooth tip, comfort eyes and funnel end. Radiopaque.
Catalog No. 50-283-0799 Supplier Med Vet International Supplier No. CATH12P
Clear Vinyl Intermittent Self Catheters Suitable for male or female catheterization. Smooth tip, comfort eyes and funnel end. Radiopaque.