Learn More
Med Vet International Red Rubber Urethral Catheter 14Fr, Sterile, 12/Case
Catalog No. 502832466
Encompass_Preferred
Red Rubber Latex Urethral Catheters: Suitable For Male Or Female Catheterization. Smooth Tip, Two Oval Eyes And Funnel End. 16" Length And Radiopaque. 14Fr.
Catalog No. 50-283-2466 Supplier Med Vet International Supplier No. DYND13514
Red Rubber Latex Urethral Catheters: Suitable For Male Or Female Catheterization. Smooth Tip, Two Oval Eyes And Funnel End. 16" Length And Radiopaque. 14Fr.