Learn More
Med Vet International Urethral Rubber Catheter, 18Fr, Sterile, Red, Each
Catalog No. 502522429
Encompass_Preferred
Red Rubber Latex Uretheral Catheter. Suitable for male or female catheterization. Smooth tip, two oval eyes and funnel end. 16" length and radiopaque. sterile.
Catalog No. 50-252-2429 Supplier Med Vet International Supplier No. CATH18R
Red Rubber Latex Uretheral Catheter. Suitable for male or female catheterization. Smooth tip, two oval eyes and funnel end. 16" length and radiopaque. sterile.