Learn More
Medchemexpress LLC HY-N0252 100mg Medchemexpress, Catharanthine CAS:2468-21-5 Purity:>98%
SDP
Catalog No. 501970242
Encompass_Preferred
Medchemexpress, HY-N0252 100mg Catharanthine CAS:2468-21-5 Catharanthine is an alkaloid isolated from Madagascar periwinkle, inhibits voltage-operated L-type Ca2+ channel, with anti-cancer and blood pressure-lowering activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0242 Supplier Medchemexpress LLC Supplier No. HYN0252100MG
Medchemexpress, HY-N0252 100mg Catharanthine CAS:2468-21-5 Catharanthine is an alkaloid isolated from Madagascar periwinkle, inhibits voltage-operated L-type Ca2+ channel, with anti-cancer and blood pressure-lowering activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.