Learn More
Apexbio Technology LLC CATHARANTHINE-SULFATE-50MG
SDP
Catalog No. 5000567495
Encompass_Preferred
Catharanthine Sulfate (CAS No 70674-90-7) is a prominent compound in biomedical research primarily used in the study of alkaloid mechanisms and therapeutic applications Originating from the plant Catharanthus roseus it plays a crucial role in understanding the biosynthesis of vinblastine and other vinca alkaloids The mechanism of action involves interactions with microtubule dynamics making it a valuable tool in cancer research In vitro its activity is observed in the nanomolar to low micromolar range Catharanthine Sulfate is frequently utilized in various cell models and animal studies for drug screening and exploring chemotherapeutic potentials Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67495 Supplier Apexbio Technology LLC Supplier No. N205350
Catharanthine Sulfate (CAS No 70674-90-7) is a prominent compound in biomedical research primarily used in the study of alkaloid mechanisms and therapeutic applications Originating from the plant Catharanthus roseus it plays a crucial role in understanding the biosynthesis of vinblastine and other vinca alkaloids The mechanism of action involves interactions with microtubule dynamics making it a valuable tool in cancer research In vitro its activity is observed in the nanomolar to low micromolar range Catharanthine Sulfate is frequently utilized in various cell models and animal studies for drug screening and exploring chemotherapeutic potentials Experimental concentrations typically depend on the specific research design