Learn More
Enzo Life Sciences Catharanthine . tartrate (100mg). CAS: 4168-17-6
SDP
Supplier:  Enzo Life Sciences ALX350101M100
Encompass_Preferred
Starting material for the synthesis of the antitumor drugs vinblastine and vincristine. It is less active as an inhibitor of tubulin self-assembly into microtubules than the latter two compounds. Alternative name: 3,4-Didehydroibogamine-18-carboxylic acid methyl ester . tartrate, 7-Ethyl-9,10,12,13-tetrahydro-6,9-methano-5H-pyrido[1',2':1,2]azepino[4,5-b]indole-6(6aH)-carboxylic acid methyl ester . tartrate. Purity: ≥90% (assay on dried basis). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8732