Learn More
Cayman Chemical Catharanthine (tartrate), Purity: ≥98%, CAS: 4168-17-6, 50 mg
SDP
Catalog No. 502952363
Encompass_Preferred
An indole alkaloid with diverse biological activities; a precursor in the biosynthesis of the vinblastine and vincristine; inhibits microtubule assembly at 320 µM; inhibits proliferation of HCT116 cells (IC50 = 60 µg/ml); induces the release of amylase from mouse pancreas preparations at 100 µM; decreases blood pressure and heart rate in anesthetized normotensive rats at 0.5-20 mg/kg
Catalog No. 50-295-2363 Supplier Cayman Chemical Supplier No. 1169550 MG
An indole alkaloid with diverse biological activities; a precursor in the biosynthesis of the vinblastine and vincristine; inhibits microtubule assembly at 320 µM; inhibits proliferation of HCT116 cells (IC50 = 60 µg/ml); induces the release of amylase from mouse pancreas preparations at 100 µM; decreases blood pressure and heart rate in anesthetized normotensive rats at 0.5-20 mg/kg