Learn More
Cayman Chemical Catharnthus VIndolIn 50mg
SDP
Catalog No. 502952315
Encompass_Preferred
A plant alkaloid used as starting material for the synthesis of the anticancer antibiotics vinblastine and vincristine
Catalog No. 50-295-2315 Supplier Cayman Chemical Supplier No. 1176550 MG
A plant alkaloid used as starting material for the synthesis of the anticancer antibiotics vinblastine and vincristine