Learn More
Medchemexpress LLC Cathepsin A Human 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7745A50UG
Encompass_Preferred
Cathepsin A protein is an important protective factor for -galactosidase and neuraminidase maintaining their stability and optimizing enzyme activity Its carboxypeptidase activity expands its functional repertoire and can deamid tachykinins and participate in a variety of biochemical processes Cathepsin A Protein Human (HEK293 His) is the recombinant human-derived Cathepsin A expressed by HEK293 with C-6 His labeled tag The total length of Cathepsin A Protein Human (HEK293 His) is 452 a a
Catalog No. 50-001-87170