Learn More
Medchemexpress LLC Cathepsin A Human 5ug | 5ug
SDP
Catalog No. 5000187010
Encompass_Preferred
Cathepsin A protein is an important protective factor for -galactosidase and neuraminidase maintaining their stability and optimizing enzyme activity Its carboxypeptidase activity expands its functional repertoire and can deamid tachykinins and participate in a variety of biochemical processes Cathepsin A Protein Human (HEK293 His) is the recombinant human-derived Cathepsin A expressed by HEK293 with C-6 His labeled tag The total length of Cathepsin A Protein Human (HEK293 His) is 452 a a
Catalog No. 50-001-87010 Supplier Medchemexpress LLC Supplier No. HYP7745A5UG
Cathepsin A protein is an important protective factor for -galactosidase and neuraminidase maintaining their stability and optimizing enzyme activity Its carboxypeptidase activity expands its functional repertoire and can deamid tachykinins and participate in a variety of biochemical processes Cathepsin A Protein Human (HEK293 His) is the recombinant human-derived Cathepsin A expressed by HEK293 with C-6 His labeled tag The total length of Cathepsin A Protein Human (HEK293 His) is 452 a a