Learn More
Medchemexpress LLC Cathepsin A Mouse 10ug
SDP
Catalog No. 5000178556
Encompass_Preferred
Cathepsin A a multifunctional protein crucially supports beta-galactosidase and neuraminidase activities It forms protective associations with these enzymes ensuring their stability and overall function Cathepsin A also displays carboxypeptidase activity and the ability to deamidate tachykinins highlighting its diverse enzymatic functions Cathepsin A Protein Mouse (HEK293 His) is the recombinant mouse-derived Cathepsin A protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-78556 Supplier Medchemexpress LLC Supplier No. HYP7434610UG
Cathepsin A a multifunctional protein crucially supports beta-galactosidase and neuraminidase activities It forms protective associations with these enzymes ensuring their stability and overall function Cathepsin A also displays carboxypeptidase activity and the ability to deamidate tachykinins highlighting its diverse enzymatic functions Cathepsin A Protein Mouse (HEK293 His) is the recombinant mouse-derived Cathepsin A protein expressed by HEK293 with C-10 His labeled tag