Learn More
Sigma Aldrich Fine Chemicals Biosciences Cathepsin B Active human r
Catalog No. 502905529
Encompass
Human cathepsin B (GenBank Accession No. NM 001908) CD33 signal peptide(amino acids 1-16) Cathepsin B (amino acids 18-339) with C-terminal HIS tag MW 43 kDa expressed in freeStyle 293-f cells. Cathepsin B is a cysteine protease found in the lysosome of normal cells and tissues. Cathepsin B active human is encoded by the gene CTSB located on human chromosome 8p23.1.
Catalog No. 50-290-5529 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP028910UG
Add to Cart
Add to Cart
Human cathepsin B (GenBank Accession No. NM 001908) CD33 signal peptide(amino acids 1-16) Cathepsin B (amino acids 18-339) with C-terminal HIS tag MW 43 kDa expressed in freeStyle 293-f cells. Cathepsin B is a cysteine protease found in the lysosome of normal cells and tissues. Cathepsin B active human is encoded by the gene CTSB located on human chromosome 8p23.1.