Learn More
Cayman Chemical CathepsIn BhumnrecombIna 50ug
SDP
Catalog No. 502945266
Encompass_Preferred
Source: Active recombinant human C-Terminal His-tagged cathepsin B expressed in HEK293 cells • Amino acids: 74-339 • MW: 37.2 kDa
Catalog No. 50-294-5266 Supplier Cayman Chemical Supplier No. 4107150 UG
Source: Active recombinant human C-Terminal His-tagged cathepsin B expressed in HEK293 cells • Amino acids: 74-339 • MW: 37.2 kDa