Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cathepsin C/DPPI protein, mouse | 9032-68-2 | >95.0% | 100UG
SDP

Catalog No. 5000174225
Encompass_Preferred

Recombinant active mouse Cathepsin C (dipeptidyl peptidase I) expressed in HEK293 cells and supplied as a stabilized, filtered solution for biochemical assays requiring active, purified enzyme.

  • Expressed in HEK293 cells with C-terminal His tag.
  • Demonstrated activity >60,000 pmol/min/μg using GR-AMC substrate.
  • Supplied in 20 mM MES, 150 mM NaCl, pH 5.5, 20% glycerol.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass 51.14 kDa (D25-L462).
  • Store at -80°C; avoid repeated freeze-thaw; stable at -20°C for 3 months after opening.

Catalog No. 50-001-74225 Supplier Medchemexpress LLC Supplier No. HYP704018100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant active mouse Cathepsin C (dipeptidyl peptidase I) expressed in HEK293 cells and supplied as a stabilized, filtered solution for biochemical assays requiring active, purified enzyme.

  • Expressed in HEK293 cells with C-terminal His tag.
  • Demonstrated activity >60,000 pmol/min/μg using GR-AMC substrate.
  • Supplied in 20 mM MES, 150 mM NaCl, pH 5.5, 20% glycerol.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass 51.14 kDa (D25-L462).
  • Store at -80°C; avoid repeated freeze-thaw; stable at -20°C for 3 months after opening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.