Learn More
Medchemexpress LLC Cathepsin C/DPPI protein, mouse (active, HEK293, His) | 9032-68-2 | >95.0% | 10UG
SDP
Catalog No. 5000174256
Encompass_Preferred
Cathepsin C/DPPI protein, mouse (active, HEK293, His) is a recombinant, enzymatically active lysosomal cysteine protease produced in HEK293 cells, purified with a C-terminal His tag, and supplied as a filtered solution formulated for biochemical assays and substrate cleavage studies.
  • Recombinant mouse cathepsin c expressed in HEK293 cells.
  • Enzymatically active; specific activity >60,000 pmol/min/μg (GR-AMC substrate).
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Glycosylated, processed fragments consistent with native protein.
  • Formulated in 20 mM MES, 150 mM NaCl, pH 5.5, 20% glycerol; 0.22 μm filtered solution.
  • Low endotoxin level (<1 EU/μg) by LAL.
  • Available in multiple microgram sizes; ship on dry ice and store at -80°C.
Catalog No. 50-001-74256 Supplier Medchemexpress LLC Supplier No. HYP70401810UG
Cathepsin C/DPPI protein, mouse (active, HEK293, His) is a recombinant, enzymatically active lysosomal cysteine protease produced in HEK293 cells, purified with a C-terminal His tag, and supplied as a filtered solution formulated for biochemical assays and substrate cleavage studies.
  • Recombinant mouse cathepsin c expressed in HEK293 cells.
  • Enzymatically active; specific activity >60,000 pmol/min/μg (GR-AMC substrate).
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Glycosylated, processed fragments consistent with native protein.
  • Formulated in 20 mM MES, 150 mM NaCl, pH 5.5, 20% glycerol; 0.22 μm filtered solution.
  • Low endotoxin level (<1 EU/μg) by LAL.
  • Available in multiple microgram sizes; ship on dry ice and store at -80°C.