Learn More
Medchemexpress LLC Cathepsin C/DPPI protein, mouse (inactive, His-Myc) | >95.0% | 20UG
SDP
Catalog No. 5000176418
Encompass_Preferred
Recombinant mouse Cathepsin C/DPPI protein (inactive) provided as a lyophilized, His-Myc-tagged construct for research use. The protein is expressed in E. coli, supplied at high purity, formulated for stability with trehalose and buffer salts, and has low endotoxin. Storage and reconstitution recommendations are provided to preserve protein integrity and activity state.
Catalog No. 50-001-76418 Supplier Medchemexpress LLC Supplier No. HYP70124720UG
Recombinant mouse Cathepsin C/DPPI protein (inactive) provided as a lyophilized, His-Myc-tagged construct for research use. The protein is expressed in E. coli, supplied at high purity, formulated for stability with trehalose and buffer salts, and has low endotoxin. Storage and reconstitution recommendations are provided to preserve protein integrity and activity state.