Learn More
Medchemexpress LLC Cathepsin C/DPPI protein, mouse (inactive, His-Myc) | >95.0% | 250UG
SDP
Supplier:  Medchemexpress LLC HYP701247250UG
Encompass_Preferred
Recombinant inactive mouse Cathepsin C/DPPI protein fragment expressed in E. coli and supplied lyophilized with N-terminal 10xHis and C-terminal Myc tags. The product corresponds to residues D25-W134, is provided at high purity, and is intended for biochemical, structural, and antibody development applications.
  • Inactive recombinant mouse Cathepsin C fragment for nonenzymatic studies.
  • N-terminal 10xHis and C-terminal Myc tags for detection and purification.
  • >95.0% purity by reducing SDS-PAGE.
  • Lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0).
  • Stable when stored lyophilized at -20°C; reconstituted stability: 4°C for 1 week or -20°C with carrier.
  • Approximate molecular weight 16-20 kDa.
Catalog No. 50-001-76440