Learn More
Diagnostic Biosystems CATHEPSIN D Cathepsin D
Catalog No. 502407471
Encompass_Preferred
Cathepsin D is an estrogen-regulated protein associated with tissue breakdown. Levels of cathepsin D have been positively correlated with recurring breast cancers of both node-negative and node-positive types. Additionally cathepsin D has been associated with amyloid formation in Alzheimer's plaques. As cathepsin D activity is increased by cigarette smoke, the enzyme may contribute to lung tissue damage in smokers.
Catalog No. 50-240-7471 Supplier Diagnostic Biosystems Supplier No. PDR004
Cathepsin D is an estrogen-regulated protein associated with tissue breakdown. Levels of cathepsin D have been positively correlated with recurring breast cancers of both node-negative and node-positive types. Additionally cathepsin D has been associated with amyloid formation in Alzheimer's plaques. As cathepsin D activity is increased by cigarette smoke, the enzyme may contribute to lung tissue damage in smokers.