Learn More
Medchemexpress LLC Cathepsin D Cricetu 5ug
SDP
Catalog No. 5000183219
Encompass_Preferred
Cathepsin D Protein is a recombinant lysosomal aspartic protease derived from Cricetulus griseus and supplied with a His-SUMO fusion tag to aid expression and purification. It is provided as a purified research reagent for biochemical assays, enzyme characterization, and antibody generation.
  • Recombinant protein derived from Cricetulus griseus.
  • His-SUMO fusion tag for improved solubility and purification.
  • Suitable for biochemical assays and enzyme characterization.
  • High purity preparation intended for research use.
  • Documents available include datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-001-83219 Supplier Medchemexpress LLC Supplier No. HYP721555UG
Cathepsin D Protein is a recombinant lysosomal aspartic protease derived from Cricetulus griseus and supplied with a His-SUMO fusion tag to aid expression and purification. It is provided as a purified research reagent for biochemical assays, enzyme characterization, and antibody generation.
  • Recombinant protein derived from Cricetulus griseus.
  • His-SUMO fusion tag for improved solubility and purification.
  • Suitable for biochemical assays and enzyme characterization.
  • High purity preparation intended for research use.
  • Documents available include datasheet, certificate of analysis, and safety data sheet.