Learn More
Medchemexpress LLC Cathepsin D and E FRET Substrate acetate | 99.8% | 1816.15 | 10 MG
SDP
Catalog No. 5000423211
Encompass_Preferred
Cathepsin D and E FRET Substrate acetate is a fluorogenic substrate designed for cathepsins D and E, but not for cathepsins B, H, or L. Its cleavage occurs at the Phe-Phe amide bond, making it a valuable tool for routine assays and mechanistic studies concerning cathepsins E and D. This product is intended for research use only.
  • Fluorogenic substrate for cathepsins D and E
  • Cleavage occurs at the Phe-Phe amide bond
  • Valuable tool for routine assays
  • Useful for mechanistic studies on cathepsins E and D
Catalog No. 50-004-23211 Supplier Medchemexpress LLC Supplier No. HYP2498A10MG
Cathepsin D and E FRET Substrate acetate is a fluorogenic substrate designed for cathepsins D and E, but not for cathepsins B, H, or L. Its cleavage occurs at the Phe-Phe amide bond, making it a valuable tool for routine assays and mechanistic studies concerning cathepsins E and D. This product is intended for research use only.
  • Fluorogenic substrate for cathepsins D and E
  • Cleavage occurs at the Phe-Phe amide bond
  • Valuable tool for routine assays
  • Useful for mechanistic studies on cathepsins E and D