Learn More
Medchemexpress LLC Cathepsin D and E FRET Substrate acetate | 99.8% | 1816.15 | 25 MG
SDP
Catalog No. 5000426891
Encompass_Preferred
Cathepsin D and E FRET Substrate acetate is a fluorogenic substrate specifically for cathepsins D and E, and not for cathepsins B, H, or L. The cleavage of this substrate occurs at the Phe-Phe amide bond. It serves as a valuable tool for routine assays and mechanistic studies concerning cathepsins E and D.
  • Fluorogenic substrate
  • Specific for cathepsins D and E
  • Does not act on cathepsins B, H, or L
  • Cleavage at the Phe-Phe amide bond
  • Suitable for routine assays
  • Useful for mechanistic studies on cathepsins E and D
Catalog No. 50-004-26891 Supplier Medchemexpress LLC Supplier No. 25MGHYP2498A
Cathepsin D and E FRET Substrate acetate is a fluorogenic substrate specifically for cathepsins D and E, and not for cathepsins B, H, or L. The cleavage of this substrate occurs at the Phe-Phe amide bond. It serves as a valuable tool for routine assays and mechanistic studies concerning cathepsins E and D.
  • Fluorogenic substrate
  • Specific for cathepsins D and E
  • Does not act on cathepsins B, H, or L
  • Cleavage at the Phe-Phe amide bond
  • Suitable for routine assays
  • Useful for mechanistic studies on cathepsins E and D