Learn More
Athens Research And Technology Inc CATHEPSIN D HU LIVER 25UG
Supplier:  Athens Research And Technology Inc 161203010425UG
Encompass_Preferred
Cathepsin D, Human Liver: 25 UG; Lyophilized from 2 mM Na phosphate, pH 6.5.; MW 42,000; Extinction Coefficient=1.0; Activity: > =300 units per mg protein. Purity: > =95% by SDS-PAGE. Prepared from human tissue in the Athens Research & Tech lab.
Catalog No. 50-489-692