Learn More
Medchemexpress LLC Cathepsin D Human 50ug | 50ug
SDP
Catalog No. 5000187218
Encompass_Preferred
Cathepsin D protein is an acidic protease that actively participates in intracellular protein breakdown Studies have shown that it is critical in the processing of amyloid precursor protein (APP) with ADAM30-mediated cleavage and activation leading to APP degradation Cathepsin D Protein Human (HEK293 C-His) is the recombinant human-derived Cathepsin D protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-87218 Supplier Medchemexpress LLC Supplier No. HYP7748A50UG
Cathepsin D protein is an acidic protease that actively participates in intracellular protein breakdown Studies have shown that it is critical in the processing of amyloid precursor protein (APP) with ADAM30-mediated cleavage and activation leading to APP degradation Cathepsin D Protein Human (HEK293 C-His) is the recombinant human-derived Cathepsin D protein expressed by HEK293 with C-6 His labeled tag