Learn More
Medchemexpress LLC Cathepsin D Mouse 100ug | 100ug
SDP
Catalog No. 5000186934
Encompass_Preferred
Cathepsin D Protein Mouse (HEK293 His) is an approximately 56 0 kDa mouse cathepsin D with a His tag Cathepsin D is a lysosomal aspartic protease of the pepsin family[1]
Catalog No. 50-001-86934 Supplier Medchemexpress LLC Supplier No. HYP7749100UG
Cathepsin D Protein Mouse (HEK293 His) is an approximately 56 0 kDa mouse cathepsin D with a His tag Cathepsin D is a lysosomal aspartic protease of the pepsin family[1]