Learn More
Enzo Life Sciences Cathepsin F (human), (recombinant) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5680005
Encompass_Preferred
Highly active. MW: ~39kDa. UniProt ID: Q9UBX1. Source: Produced in insect cells (secreted) as untagged proenzyme; activated during purification. Produced in a baculovirus expression system. Purification: Purified by multi-step chromatography. Formulation: Liquid. In 20mM NaOAc, pH 5.0, containing 2.5mM EDTA, 250mM sodium chloride and 20mM L-cysteine.
Catalog No. 50-210-0549