Learn More
Enzo Life Sciences Cathepsin G (human neutrophils), (purified) (100 µg). CAS: 56645-49-9
SDP
Catalog No. 502011968
Encompass_Preferred
Purity: ≥95% by SDS-PAGE. Formulation: Lyophilized. Source: Isolated from human neutrophils. Active enzyme. Negative for HbsAg, anti-HBc, anti-HCV, and anti-HIV-1 and -2. Long Term Storage: -80°C.
Catalog No. 50-201-1968 Supplier Enzo Life Sciences Supplier No. BMLSE2830100
Purity: ≥95% by SDS-PAGE. Formulation: Lyophilized. Source: Isolated from human neutrophils. Active enzyme. Negative for HbsAg, anti-HBc, anti-HCV, and anti-HIV-1 and -2. Long Term Storage: -80°C.