Learn More
Athens Research And Technology Inc CATHEPSIN G HU NEUTROPHL 100UG
Catalog No. 50489696
Encompass_Preferred
Cathepsin G, Human Neutrophil: 100 UG; Salt-free lyophilized solid; MW 23,500; Extinction Coefficient=0.664; Activity: 2-4 units per mg protein. Purity: > =95% by SDS-PAGE.
Catalog No. 50-489-696 Supplier Athens Research And Technology Inc Supplier No. 1614030107100UG
Cathepsin G, Human Neutrophil: 100 UG; Salt-free lyophilized solid; MW 23,500; Extinction Coefficient=0.664; Activity: 2-4 units per mg protein. Purity: > =95% by SDS-PAGE.