Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cathepsin G inhibitor I | 429676-93-7 | MFCD28137715 | 98.0% | 620.63 | C36H33N2O6P | 1 ML
SDP

Catalog No. 5000244979
Encompass_Preferred

Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of the serine protease cathepsin G with reported IC50 53 nM and Ki 63 nM. The compound is supplied in solution and solid forms for use in biochemical and cellular assays requiring cathepsin G inhibition.

  • Potent inhibitor of cathepsin G (IC50 53 nM, Ki 63 nM).
  • Reversible, competitive, non-peptide mechanism of action.
  • Available as a 10 mM solution in DMSO (1 ml) or as solid in multiple pack sizes.
  • Molecular formula C36H33N2O6P and molecular weight ~620.63.
  • Suitable for biochemical and cellular inhibition assays.
  • Batch purity may vary; verify certificate of analysis before use.

Catalog No. 50-002-44979 Supplier Medchemexpress LLC Supplier No. HY10335110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of the serine protease cathepsin G with reported IC50 53 nM and Ki 63 nM. The compound is supplied in solution and solid forms for use in biochemical and cellular assays requiring cathepsin G inhibition.

  • Potent inhibitor of cathepsin G (IC50 53 nM, Ki 63 nM).
  • Reversible, competitive, non-peptide mechanism of action.
  • Available as a 10 mM solution in DMSO (1 ml) or as solid in multiple pack sizes.
  • Molecular formula C36H33N2O6P and molecular weight ~620.63.
  • Suitable for biochemical and cellular inhibition assays.
  • Batch purity may vary; verify certificate of analysis before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.