Learn More
Medchemexpress LLC Cathepsin G inhibitor I | 429676-93-7 | MFCD28137715 | ≥99.0% | 620.63 | C36H33N2O6P | 100 MG
SDP
Catalog No. 5000244731
Encompass_Preferred
Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used in biochemical and pharmacological research to probe protease activity. It demonstrates potent inhibition and is supplied as a solid with high purity suitable for assay development and mechanistic studies.
  • Selective, reversible inhibitor of cathepsin G.
  • Potent activity (IC50 53 nM; Ki 63 nM).
  • High purity (≥99.0%) for research applications.
  • Solid form, convenient for storage and handling.
  • Available in milligram-scale quantities for laboratory use.
Catalog No. 50-002-44731 Supplier Medchemexpress LLC Supplier No. HY103351100MG
Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used in biochemical and pharmacological research to probe protease activity. It demonstrates potent inhibition and is supplied as a solid with high purity suitable for assay development and mechanistic studies.
  • Selective, reversible inhibitor of cathepsin G.
  • Potent activity (IC50 53 nM; Ki 63 nM).
  • High purity (≥99.0%) for research applications.
  • Solid form, convenient for storage and handling.
  • Available in milligram-scale quantities for laboratory use.