Learn More
Medchemexpress LLC Cathepsin G inhibitor I | 429676-93-7 | MFCD28137715 | >=99.0% | 620.63 | C36H33N2O6P | 25 MG
SDP
Catalog No. 5000244555
Encompass_Preferred
Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G with nanomolar potency (IC50 = 53 nM; Ki = 63 nM). It is provided in both solid and DMSO solution formats for use in biochemical and cell-based assays to study serine protease inhibition.
  • Selective inhibition of cathepsin G (IC50 53 nM; Ki 63 nM).
  • Reversible, competitive, non-peptide mechanism suitable for biochemical assays.
  • Available in multiple solid quantities and solution formats for assay flexibility.
  • High purity (≥99.0%) for reproducible results.
  • Molecular weight 620.63 and molecular formula C36H33N2O6P.
Catalog No. 50-002-44555 Supplier Medchemexpress LLC Supplier No. HY10335125MG
Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G with nanomolar potency (IC50 = 53 nM; Ki = 63 nM). It is provided in both solid and DMSO solution formats for use in biochemical and cell-based assays to study serine protease inhibition.
  • Selective inhibition of cathepsin G (IC50 53 nM; Ki 63 nM).
  • Reversible, competitive, non-peptide mechanism suitable for biochemical assays.
  • Available in multiple solid quantities and solution formats for assay flexibility.
  • High purity (≥99.0%) for reproducible results.
  • Molecular weight 620.63 and molecular formula C36H33N2O6P.