Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cathepsin G inhibitor I | 429676-93-7 | MFCD28137715 | >99.0% | 620.63 g/mol | C36H33N2O6P | 1 MG
SDP

Catalog No. 5000244826
Encompass_Preferred

Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used for biochemical and cellular research to inhibit cathepsin G activity.

  • Selective reversible competitive inhibitor of cathepsin G (IC50 = 53 nM; Ki = 63 nM).
  • High purity solid, white to off-white, suitable for analytical and bioassay work.
  • Molecular formula C36H33N2O6P; molecular weight 620.63 g/mol.
  • Recommended storage: sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Suitable for biochemical assays and cellular studies to probe cathepsin G function.

Catalog No. 50-002-44826 Supplier Medchemexpress LLC Supplier No. HY1033511MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used for biochemical and cellular research to inhibit cathepsin G activity.

  • Selective reversible competitive inhibitor of cathepsin G (IC50 = 53 nM; Ki = 63 nM).
  • High purity solid, white to off-white, suitable for analytical and bioassay work.
  • Molecular formula C36H33N2O6P; molecular weight 620.63 g/mol.
  • Recommended storage: sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Suitable for biochemical assays and cellular studies to probe cathepsin G function.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.