Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cathepsin G inhibitor I | 429676-93-7 | MFCD28137715 | ≥99.0% | 620.63 g/mol | C36H33N2O6P | 10 MG
SDP

Catalog No. 5000244569
Encompass_Preferred

Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used in biochemical and cellular research. It is supplied as a white to off-white solid with high reported purity for use in enzymatic assays and inhibitor profiling.

  • Selective cathepsin G inhibition (IC50 = 53 nM; Ki = 63 nM)
  • Reversible, competitive, non-peptide inhibitor
  • High purity reported ≥99.0%
  • Molecular weight 620.63 g/mol
  • Molecular formula C36H33N2O6P
  • Supplied in small milligram pack sizes suitable for assay development (e.g., 10 mg)

Catalog No. 50-002-44569 Supplier Medchemexpress LLC Supplier No. HY10335110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used in biochemical and cellular research. It is supplied as a white to off-white solid with high reported purity for use in enzymatic assays and inhibitor profiling.

  • Selective cathepsin G inhibition (IC50 = 53 nM; Ki = 63 nM)
  • Reversible, competitive, non-peptide inhibitor
  • High purity reported ≥99.0%
  • Molecular weight 620.63 g/mol
  • Molecular formula C36H33N2O6P
  • Supplied in small milligram pack sizes suitable for assay development (e.g., 10 mg)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.