Learn More
Medchemexpress LLC Cathepsin inhibitor 1 | 225120-65-0 | 99.3% | 401.89 | C20H24ClN5O2 | 1 ML
SDP
Catalog No. 5000215591
Encompass_Preferred
Cathepsin inhibitor 1 is a small-molecule inhibitor of cathepsin proteases for research use. It is offered as a ready-to-use 10 mM solution in DMSO and as solid material, allowing flexible use in biochemical profiling and cellular assays. High purity and a defined molecular weight support reproducible assay performance and accurate dosing.
Catalog No. 50-002-15591 Supplier Medchemexpress LLC Supplier No. HY10023110MM 1ML
Cathepsin inhibitor 1 is a small-molecule inhibitor of cathepsin proteases for research use. It is offered as a ready-to-use 10 mM solution in DMSO and as solid material, allowing flexible use in biochemical profiling and cellular assays. High purity and a defined molecular weight support reproducible assay performance and accurate dosing.