Learn More
Medchemexpress LLC Cathepsin Inhibitor 2 | 1017931-53-1 | 421.38 | 1 ML
SDP
Catalog No. 5000449513
Encompass_Preferred
Cathepsin Inhibitor 2 is a potent Cathepsin S inhibitor, with a Ki of <20 nM. It also potently inhibits Cathepsin F, Cathepsin L, and Cathepsin B with Kis ranging from 21-500 nM.
  • Potent Cathepsin S inhibitor with Ki <20 nM.
  • Inhibits Cathepsin F, Cathepsin L, and Cathepsin B with Kis of 21-500 nM.
Catalog No. 50-004-49513 Supplier Medchemexpress LLC Supplier No. HYU0037710MM/1ML
Cathepsin Inhibitor 2 is a potent Cathepsin S inhibitor, with a Ki of <20 nM. It also potently inhibits Cathepsin F, Cathepsin L, and Cathepsin B with Kis ranging from 21-500 nM.
  • Potent Cathepsin S inhibitor with Ki <20 nM.
  • Inhibits Cathepsin F, Cathepsin L, and Cathepsin B with Kis of 21-500 nM.